Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2,4-Diaminobutyric Acid Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D117525G
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 65427-54-5
Molecular Formula C4H12Cl2N2O2
Molecular Weight (g/mol) 191.052
MDL Number MFCD00012910
InChI Key CKAAWCHIBBNLOJ-UHFFFAOYSA-N
Synonym dl-2,4-diaminobutyric acid dihydrochloride, 2,4-diaminobutanoic acid dihydrochloride, h-dl-dab.2hcl, l-2,4-diaminobutyric acid dihydrochloride, dl-2,4-diaminobutyricaciddihydrochloride, butanoic acid, 2,4-diamino-, dihydrochloride, l-2,4-diaminobutyric aicd dihydrochloride, 2,4-bis azanyl butanoic acid dihydrochloride, c4h10n2o2.2hcl, h-dl-dab-oh.2hcl
PubChem CID 2724329
IUPAC Name 2,4-diaminobutanoic acid;dihydrochloride
SMILES C(CN)C(C(=O)O)N.Cl.Cl

Specifications

Melting Point 203°C
Color White
Quantity 25 g
Formula Weight 191.05
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material DL-2,4-Diaminobutyric Acid Dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.