Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Amino-3-methyl-1-butanol, 97%, Thermo Scientific Chemicals

Catalog No. p-57383
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16369-05-4
MDL Number MFCD00004730
InChI Key NWYYWIJOWOLJNR-UHFFFAOYSA-N
Synonym 2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol
PubChem CID 79019
IUPAC Name 2-amino-3-methylbutan-1-ol
SMILES CC(C)C(CO)N

Specifications

Color Yellow
Density 0.9300g/mL
Boiling Point 75.0°C to 77.0°C (8.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H13NO
Refractive Index 1.4533 to 1.4553
Linear Formula (CH3)2CHCH(NH2)CH2OH
Quantity 5 g
Fieser 12,563; 13,341; 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,7
Specific Gravity 0.93
Molecular Weight (g/mol) 103.16
Formula Weight 103.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material DL-2-Amino-3-methyl-1-butanol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 240-425-0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.