Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Hydroxybutyric acid sodium salt, 97+%

Catalog No. AAA1863606
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 5094-24-6 | C4H7NaO3 | 126.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5094-24-6
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00004565
InChI Key MOSCXNXKSOHVSQ-UHFFFAOYNA-M
Synonym sodium 2-hydroxybutanoate, sodium 2-hydroxybutyrate, sodium dl-2-hydroxybutyrate, 2-hydroxybutyric acid sodium salt, dl-2-hydroxybutyric acid sodium salt, sodium hydroxybutyrate, acmc-1btqj, sodium, a-hydroxybutyrate, sodium 2-oxidanylbutanoate, sodiumdl-2-hydroxybutyrate
PubChem CID 23663641
IUPAC Name sodium;2-hydroxybutanoate
SMILES [Na+].CCC(O)C([O-])=O

Specifications

Melting Point 153°C to 157°C
Quantity 5 g
Beilstein 4825104
Sensitivity Hygroscopic
Formula Weight 126.09
Percent Purity ≥97%
Chemical Name or Material DL-2-Hydroxybutyric acid sodium salt
EINECSNumber 225-812-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.