Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Octanol, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
1L
2.5L
Packaging:
Glass bottle
123-96-6
C8H18O
130.23
MFCD00004591
SJWFXCIHNDVPSH-UHFFFAOYSA-N
2-octanol,2-octyl alcohol,hexylmethylcarbinol,2-hydroxyoctane,sec-caprylic alcohol,1-methylheptanol,methylhexylcarbinol,1-methylheptyl alcohol,dl-2-octanol,n-octan-2-ol
20083
CHEBI:37869
octan-2-ol
CCCCCCC(C)O
123-96-6
C8H18O
130.23
MFCD00004591
SJWFXCIHNDVPSH-UHFFFAOYSA-N
2-octanol,2-octyl alcohol,hexylmethylcarbinol,2-hydroxyoctane,sec-caprylic alcohol,1-methylheptanol,methylhexylcarbinol,1-methylheptyl alcohol,dl-2-octanol,n-octan-2-ol
20083
CHEBI:37869
octan-2-ol
CCCCCCC(C)O
Melting Point -38.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Color Colorless to Yellow
Refractive Index 1.4250 to 1.4270
Packaging Glass bottle
Boiling Point 174.0°C to 181.0°C
Density 0.8100g/mL
Quantity 2.5L
Specific Gravity 0.81
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)5CH(OH)CH3
Beilstein 01, 419
Fieser 01,109
Merck Index 15, 6841
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in most organic solvents
Formula Weight 130.23
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material DL-2-Octanol
Show More Show Less

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 204-667-