Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Pentanol, 98%, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
Packaging:
Glass bottle
6032-29-7
C5H12O
88.15
MFCD00004579
JYVLIDXNZAXMDK-UHFFFAOYSA-N
2-pentanol,sec-amyl alcohol,methylpropylcarbinol,sec-pentyl alcohol,pentanol-2,methyl propyl carbinol,2-pentyl alcohol,2-hydroxypentane,1-methyl-1-butanol,alpha-methylbutanol
22386
CHEBI:77518
pentan-2-ol
CCCC(C)O
6032-29-7
C5H12O
88.15
MFCD00004579
JYVLIDXNZAXMDK-UHFFFAOYSA-N
2-pentanol,sec-amyl alcohol,methylpropylcarbinol,sec-pentyl alcohol,pentanol-2,methyl propyl carbinol,2-pentyl alcohol,2-hydroxypentane,1-methyl-1-butanol,alpha-methylbutanol
22386
CHEBI:77518
pentan-2-ol
CCCC(C)O
Refractive Index 1.4050 to 1.407
Melting Point -50°C
Boiling Point 118°C to 119°C
Flash Point 34°C
Specific Gravity 0.812
Viscosity 4.1 mPa.s (20°C)
Infrared Spectrum Authentic
Water 0.3% max.
Quantity 500mL
Density 0.8120g/mL
Packaging Glass bottle
Residue after Evaporation 0.003% max.
Color Colorless
Assay Percent Range 98%
Linear Formula CH3CH2CH2CH(OH)CH3
Beilstein 01, 384
Merck Index 15, 7231
Solubility Information Solubility in water: very soluble. Other solubilities: miscible with alcohol and ether
Formula Weight 88.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material DL-2-Pentanol
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning