Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-3-Octanol, 97%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
Packaging:
Glass bottle
589-98-0
C8H18O
130.23
MFCD00004590
NMRPBPVERJPACX-UHFFFAOYSA-N
3-octanol,octanol-3,amyl ethyl carbinol,ethyl-n-amylcarbinol,amylethylcarbinol,ethylamylcarbinol,1-ethylhexanol,d-n-octanol,ethyl amyl carbinol,s-3-octanol
11527
CHEBI:80945
octan-3-ol
CCCCCC(CC)O
589-98-0
C8H18O
130.23
MFCD00004590
NMRPBPVERJPACX-UHFFFAOYSA-N
3-octanol,octanol-3,amyl ethyl carbinol,ethyl-n-amylcarbinol,amylethylcarbinol,ethylamylcarbinol,1-ethylhexanol,d-n-octanol,ethyl amyl carbinol,s-3-octanol
11527
CHEBI:80945
octan-3-ol
CCCCCC(CC)O
Boiling Point 174.0°C to 176.0°C
Color Colorless
Infrared Spectrum Authentic
Quantity 250mL
Refractive Index 1.4220 to 1.4280
Flash Point 67°C
Density 0.8250g/mL
Melting Point -45.0°C
Packaging Glass bottle
Specific Gravity 0.825
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)4CH(OH)C2H5
Beilstein 01, I, 208
Solubility Information Solubility in water: insoluble.
Formula Weight 130.23
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material DL-3-Octanol
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 209-667-4