Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DL-AP4
GREENER_CHOICESDP

Catalog No. 0101100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-011-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-011-00 Supplier Tocris Bioscience™ Supplier No. 0101/100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Broad spectrum glutamatergic antagonist

Broad spectrum EAA ligand.

D-isomer, L-isomer and Sodium Salt also available.

Chemical Identifiers

CAS 20263-07-4
Molecular Formula C4H10NO5P

Specifications

Quantity 100 mg
Formula Weight 183.1
Purity >98%
Molecular Weight (g/mol) 183.10
Chemical Name or Material DL-AP4
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.