Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DL-AP5 Sodium salt
GREENER_CHOICE

Catalog No. 369310R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-931-0R 10 mg
36-935-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-931-0R Supplier Tocris Bioscience™ Supplier No. 3693/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NMDA antagonist; sodium salt of DL-AP5

DL-AP5 Sodium salt is the sodium salt of DL-AP5 (Cat. No. 0105) and a racemic mixture of the D- and L-isomers of AP5, a selective NMDA receptor antagonist that competes with glutamate binding and is commonly used to inhibit NMDA-dependent synaptic plasticity. D-AP5 (Cat. No. 0106) is the more active isomer and displays approximately 52-fold higher potency than the L-isomer, L-AP5 (Cat. No. 0107). In vitro D-AP5 reduces NMDA-induced depolarization of cortical neurons, with no effect on the response to L-Quisqualic acid (Cat. No. 0188) or Kainic acid (Cat. No. 0222). Following spinal injection of D-AP5, NMDA-response is rapidly reduced, with no effect seen on spontaneously active neurons.

DL mixture, D-isomer and L-isomer also available.

Specifications

Quantity 10 mg
Product Type DL-AP5 Sodium salt
Purity >98%
Molecular Weight (g/mol) 219.11
Inhibitors DL-AP5 Sodium salt

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.