Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Asparagine Monohydrate 99.0+%, TCI America™
SDP

Catalog No. A0541100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0541100G 100 g
A054125G 25 g
A0541500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0541100G Supplier TCI America Supplier No. A0541100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3130-87-8
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00151039,MFCD00008036,MFCD00064400
InChI Key DCXYFEDJOCDNAF-UHFFFAOYNA-N
Synonym dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
PubChem CID 522362
IUPAC Name 2-amino-3-carbamoylpropanoic acid
SMILES NC(CC(N)=O)C(O)=O

Specifications

Color White
Quantity 100 g
Formula Weight 132.12
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Asparagine Monohydrate
EINECSNumber (2)-2383
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.