Learn More
DL-beta-Proline hydrochloride, 95%
Description
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Specifications
| Melting Point | 30°C to 38°C |
| Quantity | 1 g |
| Flash Point | >110°C (230°F) |
| Formula Weight | 151.59 |
| Percent Purity | 95% |
| Chemical Name or Material | DL-beta-Proline hydrochloride |
Safety and Handling
| Hazard Category | H318 |
| Hazard Statement | GHS H Statement H318 Causes serious eye damage. |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.