Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Cysteine, 96%

Catalog No. p-7037402
Click to view available options
Quantity:
5 g
25 g

CAS: 3374-22-9 | C3H7NO2S | 121.15 g/mol

DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

Solubility
Soluble in water (partly), 1 N HCl (25 mg/ml), ethanol, and acetic acid. Insoluble in acetone.

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3374-22-9
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.15
MDL Number MFCD00004881
InChI Key XUJNEKJLAYXESH-UHFFFAOYNA-N
Synonym dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh
PubChem CID 594
ChEBI CHEBI:15356
IUPAC Name 2-amino-3-sulfanylpropanoic acid
SMILES NC(CS)C(O)=O

Specifications

Melting Point 225°C (decomposition)
Beilstein 1721406
Solubility Information Soluble in water (partly),1 N HCl (25mg/ml),ethanol,and acetic acid. Insoluble in acetone.
Quantity 5 g
Formula Weight 121.16
Percent Purity 96%
Chemical Name or Material DL-Cysteine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 222-160-2
RTECSNumber HA1595000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.