Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Cystine, 99%, Thermo Scientific™

Catalog No. AC303030025
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923-32-0
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00084652
InChI Key LEVWYRKDKASIDU-UHFFFAOYSA-N
Synonym dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l
PubChem CID 595
ChEBI CHEBI:17376
IUPAC Name 2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Color White
Chlorine (Cl) 500ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 20ppm max.
Assay Percent Range 99%
Loss on Drying 0.2% max.
Packaging Glass Bottle
Linear Formula [-SCH2CH(NH2)CO2H]2
Sulfate 200ppm max.
Merck Index 15, 2779
Transmittance 95.0% min.
Quantity 2.5 g
Formula Weight 240.3
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material DL-Cystine, 99%
Show More Show Less
missing translation for 'healthHazard1' Warning
missing translation for 'healthHazard2' GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.