Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Glyceraldehyde dimer, 95%, Thermo Scientific Chemicals

Catalog No. AC450710050
Click to view available options
Quantity:
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23147-59-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key HQAMJDIBKAKYMU-UHFFFAOYSA-N
Synonym 3,6-bis hydroxymethyl-1,4-dioxane-2,5-diol, pubchem10426, dl-glyceraldehyde dimer+, 2,5-dihydroxy-3,6-dihydroxymethyl-1,4-dioxane
PubChem CID 251615
IUPAC Name 3,6-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
SMILES C(C1C(OC(C(O1)O)CO)O)O

Specifications

Melting Point 138.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 180.16
Percent Purity 95%
Chemical Name or Material DL-Glyceraldehyde dimer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.