Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich DL-Lactic acid

Catalog No. 697851L Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L

Catalog No. 69-785-1L

Supplier: Sigma Aldrich 697851L

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DL-Lactic acid can be used to synthesize serine racemase (SR) inhibitors.

Specifications

CAS 50-21-5
Boiling Point 122°C (15 mmHg)
Molecular Formula C3H6O3
Linear Formula CH3CH(OH)COOH
MDL Number MFCD00004520
Synonym 2-Hydroxypropionic acid
Molecular Weight (g/mol) 90.08
Percent Purity ∼90% (T)
Density 1.209 g/mL (at 25°C (literature))
Quantity 1 L
Refractive Index n20/D 1.425 (literature)
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.