Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Lactic acid, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
50-21-5,7732-18-5
C3H6O3
90.078
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYSA-N
lactic acid,dl-lactic acid,2-hydroxypropionic acid,milk acid,lactate,polylactic acid,ethylidenelactic acid,lactovagan,tonsillosan,acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(C(=O)O)O
50-21-5,7732-18-5
C3H6O3
90.078
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYSA-N
lactic acid,dl-lactic acid,2-hydroxypropionic acid,milk acid,lactate,polylactic acid,ethylidenelactic acid,lactovagan,tonsillosan,acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(C(=O)O)O
Acidity Citric, oxalic, phosphoric or tartaric acid: Passes Test
Assay Percent Range 85%
Linear Formula CH3CH(OH)CO2H
Merck Index 15, 5384
Density 1.2000g/mL
Formula Weight 90.08
Physical Form Liquid
Percent Purity 90%
Heavy Metals (as Pb) 10ppm max.
Identification Passes Test
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Packaging Glass bottle
Refractive Index 1.4252 to 1.4272
Residue after Ignition 0.1% max.
Specific Gravity 1.2
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol, furfurol, glycerol, less soluble in ether, practically insoluble in chloroform, carbon, disulfide, petroleum ether
Specific Rotation − 0.05
Sugars Passes Test
Specific Rotation Condition − 0.05 (25.00°C c= neat)
Melting Point 18.0°C
Color Colorless to Yellow
Boiling Point 122.0°C (15.0 mmHg)
Quantity 2.5kg
Flash Point >110°C
Chemical Name or Material DL-Lactic acid
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger