Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Mandelic acid, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
Packaging:
Plastic Bucket
Plastic bottle
90-64-2
C8H8O3
152.149
IWYDHOAUDWTVEP-UHFFFAOYSA-N
mandelic acid,dl-mandelic acid,phenylglycolic acid,amygdalic acid,almond acid,uromaline,p-mandelic acid,racemic mandelic acid,paramandelic acid,rs-mandelic acid
1292
CHEBI:35825
2-hydroxy-2-phenylacetic acid
C1=CC=C(C=C1)C(C(=O)O)O
90-64-2
C8H8O3
152.149
IWYDHOAUDWTVEP-UHFFFAOYSA-N
mandelic acid,dl-mandelic acid,phenylglycolic acid,amygdalic acid,almond acid,uromaline,p-mandelic acid,racemic mandelic acid,paramandelic acid,rs-mandelic acid
1292
CHEBI:35825
2-hydroxy-2-phenylacetic acid
C1=CC=C(C=C1)C(C(=O)O)O
Assay Percent Range 99+%
Linear Formula C6H5CH(OH)CO2H
Formula Weight 152.15
Physical Form Crystals or Crystalline Powder
Percent Purity ≥99.0% (Acid)
Infrared Spectrum Authentic
Packaging Plastic bottle
Melting Point 117°C to 121°C
Color White
Quantity 100g
Chemical Name or Material DL-Mandelic acid, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-007-6

RTECSNumber : OO6300000

TSCA : TSCA