Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Mandelic acid, 99+%

Catalog No. p-154732
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle

CAS: 90-64-2 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-64-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key IWYDHOAUDWTVEP-UHFFFAOYSA-N
Synonym mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid
PubChem CID 1292
ChEBI CHEBI:35825
IUPAC Name 2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

Melting Point 117°C to 121°C
Color White
Assay Percent Range 99+%
Linear Formula C6H5CH(OH)CO2H
Formula Weight 152.15
Percent Purity ≥99.0% (Acid)
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystals or Crystalline Powder
Chemical Name or Material DL-Mandelic acid, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-007-6
RTECSNumber OO6300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.