Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DL-Menthol, Crystal, FCC, Spectrum™ Chemical

Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical DL-Menthol, Crystal, FCC is used as a flavor and fragrance additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Boiling Point | 212°C |
Melting Range | 31°C to 44°C |
Packaging | Amber Glass Bottle |
Quantity | 2.5 kg |
Residue after Evaporation | 0.05% |
Specific Rotation | -2° to +2° |
Grade | FCC |
Physical Form | Low Melting Crystals or Crystalline Mass |
Chemical Name or Material | DL-Menthol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction