Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-α-(Methylaminomethyl)benzyl alcohol, 99%, Thermo Scientific™

Catalog No. p-159798
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6589-55-5
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00004506
InChI Key ZCTYHONEGJTYQV-UHFFFAOYSA-N
Synonym 2-methylamino-1-phenylethanol, halostachine, n-methylphenylethanolamine, +--halostachine, 2-methylamino-1-phenylethan-1-ol, +--alpha-methylamino methyl benzenemethanol, alpha-methylamino methyl benzyl alcohol, ethanol, 2-methylamino-1-phenyl, benzyl alcohol, alpha-methylamino methyl-, dl, alpha-methylaminomethyl benzyl alcohol
PubChem CID 913
ChEBI CHEBI:16913
IUPAC Name 2-(methylamino)-1-phenylethanol
SMILES CNCC(C1=CC=CC=C1)O

Specifications

Melting Point 74.0°C to 76.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula C6H5CH(CH2NHCH3)OH
Quantity 1 g
Merck Index 10, 4485
Formula Weight 151.21
Percent Purity 99%
Physical Form Fine Crystalline Needles
Chemical Name or Material DL-α-(Methylaminomethyl)benzyl alcohol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber DA4976000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.