Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-N-Benzoyl-2-methylserine, 98%, Thermo Scientific™

Catalog No. AC339790010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7508-82-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD01863306
InChI Key FVWQRKJTTRAXJP-NSHDSACASA-N
Synonym s-2-benzamido-3-hydroxy-2-methylpropanoic acid, n-benzoyl-2-methylserine, 2s-2-benzamido-3-hydroxy-2-methylpropanoic acid, s-2-methyl-2-benzoylamino-3-hydroxypropanoic acid, 2s-3-hydroxy-2-methyl-2-phenylformamido propanoic acid
PubChem CID 7167581
IUPAC Name (2S)-2-benzamido-3-hydroxy-2-methylpropanoic acid
SMILES CC(CO)(C(=O)O)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 153.0°C to 159.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol,aq. naoh,aq. na2co3
Formula Weight 223.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-N-Benzoyl-2-methylserine, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.