Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-N-FMOC-2'-Methylphenylalanine, 97%, Thermo Scientific™

Catalog No. AC326090010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135944-06-8
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD01320674
InChI Key GYFMRMRGTNDDAT-UHFFFAOYSA-N
Synonym acmc-20akv3, 2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylphenyl propanoic acid, dl-n-fmoc-2'-methylphenylalanine, 2-fluoren-9-ylmethoxy carbonylamino-3-2-methylphenyl propanoic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-methylphenyl propanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-methylphenyl propanoic acid
PubChem CID 2733445
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-methylphenyl)propanoic acid
SMILES CC1=CC=CC=C1CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 157°C to 164°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 1 g
Formula Weight 401
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material DL-N-FMOC-2'-Methylphenylalanine, 97%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.