Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ DL-Panthenol, 99%

Click to view available options
Quantity:
50g
250g
1000g
16485-10-2
C9H19NO4
205.254
MFCD00002944
SNPLKNRPJHDVJA-UHFFFAOYSA-N
dl-panthenol,panthenol,dl-pantothenol,dl-pantothenyl alcohol,panthenolum,2,4-dihydroxy-n-3-hydroxypropyl-3,3-dimethylbutanamide,pantenol,pantenolo,penthenol,varitan
4678
2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
CC(C)(CO)C(C(=O)NCCCO)O
16485-10-2
C9H19NO4
205.254
MFCD00002944
SNPLKNRPJHDVJA-UHFFFAOYSA-N
dl-panthenol,panthenol,dl-pantothenol,dl-pantothenyl alcohol,panthenolum,2,4-dihydroxy-n-3-hydroxypropyl-3,3-dimethylbutanamide,pantenol,pantenolo,penthenol,varitan
4678
2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
CC(C)(CO)C(C(=O)NCCCO)O
Applications
DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.

Solubility
Soluble in water, ethanol, ether, chloroform, and propylene glycol.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
Melting Point 66°C to 69°C
Quantity 50g
Merck Index 14,2947
Formula Weight 205.26
Percent Purity 99%
Chemical Name or Material DL-Panthenol

RTECSNumber : ES4316500

TSCA : TSCA

Recommended Storage : 2° to 8°C