Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Panthenol, 99%

Catalog No. p-7028007
Click to view available options
Quantity:
50 g
250 g

CAS: 16485-10-2 | C9H19NO4 | 205.25 g/mol

DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.

Solubility
Soluble in water, ethanol, ether, chloroform, and propylene glycol.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16485-10-2
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00002944
InChI Key SNPLKNRPJHDVJA-UHFFFAOYNA-N
Synonym dl-panthenol, panthenol, dl-pantothenol, dl-pantothenyl alcohol, panthenolum, 2,4-dihydroxy-n-3-hydroxypropyl-3,3-dimethylbutanamide, pantenol, pantenolo, penthenol, varitan
PubChem CID 4678
IUPAC Name 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)C(O)C(=O)NCCCO

Specifications

Melting Point 66°C to 69°C
Quantity 50 g
Sensitivity Hygroscopic
Merck Index 14,2947
Solubility Information Soluble in water,ethanol,ether,chloroform,and propylene glycol.
Formula Weight 205.26
Percent Purity 99%
Chemical Name or Material DL-Panthenol
EINECSNumber 240-540-6
RTECSNumber ES4316500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.