Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Pantolactone, 97%

Catalog No. p-3771069
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 79-50-5 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-50-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00064333
InChI Key SERHXTVXHNVDKA-UHFFFAOYSA-N
Synonym pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
PubChem CID 989
IUPAC Name 3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 74.0°C to 78.0°C
Boiling Point 246.0°C
Flash Point 122°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material DL-Pantolactone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 201-210-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.