Learn More
Thermo Scientific Chemicals DL-Phenylalanine, 99%
Description
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.
Solubility
Soluble in water. Insoluble in ethyl ether, benzene.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | ∼266°C (decomposition) |
Beilstein | 1910407 |
Merck Index | 14,7271 |
Solubility Information | Soluble in water. Insoluble in ethyl ether,benzene. |
Quantity | 100 g |
Formula Weight | 165.19 |
Percent Purity | 99% |
Chemical Name or Material | DL-Phenylalanine |
Safety and Handling
EINECSNumber | 205-756-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.