Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium dicyanamide, 97%, pure

Catalog No. AC176340250
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle

Catalog No. AC176340250

Supplier: Thermo Scientific Chemicals 176340250

Only null left
Add to Cart

CAS: 1934-75-4 | C2N3Na | 89.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1934-75-4
Molecular Formula C2N3Na
Molecular Weight (g/mol) 89.03
MDL Number MFCD00003535
InChI Key IXBPPZBJIFNGJJ-UHFFFAOYSA-N
Synonym sodium dicyanamide, sodium dicyanoazanide, sodium dicyanoamide, unii-i9c96i1mre, cyanamide, cyano-, sodium salt, cyanamide,cyano-,sodium salt, i9c96i1mre, cyanamide, n-cyano-, sodium salt 1:1, dicyanamide, sodium salt, nadca
PubChem CID 4350929
SMILES [Na+].N#C[N-]C#N

Specifications

Melting Point >300°C
Color Pink or White
Infrared Spectrum Authentic
Assay Percent Range 2% max. (NaCl) (Argentometry)
Packaging Plastic bottle
Linear Formula NaN(CN)2
Quantity 25 g
Beilstein 03, III, 154
Solubility Information Solubility in water: 260g/L (30°C). Other solubilities: insoluble in aceton,etoh and ethyl acetate
IUPAC Name sodium;dicyanoazanide
Formula Weight 89.03
Percent Purity 97%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium dicyanamide, Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens
EINECSNumber 217-703-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.