Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Individual Instruction: Science In The Kitchen
SCI_ED

Catalog No. S01994
Click to view available options
Class Size:
1 Lab Group

This comprehensive kit incorporates a variety of scientific techniques all themed around common kitchen materials.

This comprehensive kit incorporates a variety of scientific techniques all themed around common kitchen materials. Students will perform chromatography on commercial food colors, use chemical tests to identify an unknown cooking ingredient, examine the protein digesting ability of a common meat tenderizing enzyme, use titration to quantify vitamin C levels and then test an unknown juice or soda (not provided), and lastly examine some of the differences and similarities in the materials used to clean up (soap, hand dishwashing detergent, and machine dishwashing detergent).

Kit includes:

  • Ascorbic Acid Capsule
  • Unknown Powder, 25 g
  • Blue Food Dye, 5 mL
  • Green Food Dye, 5 mL
  • Red Food Dye, 5 mL
  • Yellow Food Dye, 5 mL
  • Gelatin Powder, 5 g
  • Baking Powder, 5 g
  • Papain Powder, 5 g
  • Baking Soda, 5 g
  • Corn Starch, 5 g
  • Vinegar, 5 mL
  • 5% Calcium Chloride, 5 mL
  • Dishwashing Detergent - Machine, 5 mL
  • Dishwashing Detergent - Hand, 5 mL
  • Iodine Potassium Iodide, 30 mL
  • Dilute Lugols Solution, 30 mL
  • 5% Soap, 10 mL
  • Starch Indicator Solution, 5 mL
  • Chromatography Paper
  • 4 Capillary Tubes
  • Centrifuge Tube
  • Spot Plate
  • Toothpicks, Package
  • 6 Medicine Cups
  • Pipette
  • Universal Indicator pH Strips

Specifications

Product Type Testing Food for Nutrients Kit
Grade 6+
Includes Ascorbic Acid Capsule, Unknown Powder,Various Food Dyes, Gelatin Powder, Baking Powder, Papain Powder, Baking Soda, Corn Starch, Vinegar, 5% Calcium Chloride, Dishwashing Detergents, Iodine, Potassium Iodide, Dilute Lugols Solution, 5% Soap, Starch Indicator Solution, Chromatography Paper, 4 Capillary Tubes, Centrifuge Tube, Spot Plate, Toothpicks, 6 Medicine Cups, Pipette, Universal Indicator pH Strips
Class Size 1 Lab Group
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.