Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Tropic acid, 97%

Catalog No. AC140550250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 552-63-6 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-63-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004255
InChI Key JACRWUWPXAESPB-UHFFFAOYNA-N
Synonym tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid
PubChem CID 10726
ChEBI CHEBI:30765
IUPAC Name 3-hydroxy-2-phenylpropanoic acid
SMILES OCC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 117.0°C to 120.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(CH2OH)CO2H
Quantity 25 g
Beilstein 10, 261
Merck Index 15, 9958
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,practically insoluble in petroleum ether
Formula Weight 166.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material DL-Tropic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 209-020-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.