Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Tryptophanamide, 98%

Catalog No. AAH6322706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6322706 5 g
AAH63227MD 250 mg
AAH6322703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6322706 Supplier Thermo Scientific Chemicals Supplier No. H6322706

CAS: 6720-02-1 | C11H13N3O | 203.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6720-02-1
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD03826622
InChI Key JLSKPBDKNIXMBS-UHFFFAOYNA-N
PubChem CID 89366
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanamide
SMILES NC(CC1=CNC2=CC=CC=C12)C(N)=O

Specifications

Color White
Synonym 2-phenoxybiphenyl, 2-phenoxy-1,1'-biphenyl, 2-phenyloxybiphenyl, phenoxy-2-diphenyl, 1,1'-biphenyl, phenoxy, 1,1'-biphenyl, 2-phenoxy, o-diphenyl phenyl ether, 2-biphenylyl phenyl ether, ether, 2-biphenylyl phenyl, phenyl biphenyl ether
Quantity 5 g
Formula Weight 203.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material DL-Tryptophanamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.