Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-Valine, 99%

Catalog No. AAA167560E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA167560E 2500 g
AAA1675622 100 g
AAA1675636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA167560E Supplier Thermo Scientific Chemicals Supplier No. A167560E
Add to Cart

CAS: 516-06-3 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYSA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O

Specifications

Melting Point ∼296°C (sublimation)
Beilstein 506689
Merck Index 14,9909
Quantity 2500 g
Formula Weight 117.15
Percent Purity 99%
Chemical Name or Material DL-Valine
EINECSNumber 208-220-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.