Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DLC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572272
Description
DLC Polyclonal Antibody for Western Blot
Acts as a ligand for Notch receptors and is involved in somitogenesis. Can activate Notch receptors. Required in somite segmentation to keep the oscillations of neighboring presomitic mesoderm cells synchronized.
Specifications
DLC | |
Polyclonal | |
Unconjugated | |
dlc | |
bea; beamter; delC; deltaC; Delta-like protein C; dlc | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
30120 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.81 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9IAT6 | |
dlc | |
KLH conjugated synthetic peptide between 557-591 amino acids from DANRE dlc. | |
200 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction