Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLD Recombinant Rabbit Monoclonal Antibody (3J2O9)
GREENER_CHOICE

Catalog No. PIMA535680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535680 Supplier Invitrogen™ Supplier No. MA535680
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: WVGKSEEQLK EEGIEYKVGK FPFAANSRAK TNADTDGMVK ILGQKSTDRV LGAHILGPGA GEMVNEAALA LEYGASCEDI ARVCHAHPTL SEAFREANLA.

This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLD
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3J2O9
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dld
Gene Accession No. O08749, P09622, Q6P6R2
Gene Alias aei; after eight protein; branched chain alpha-keto acid dehydrogenase complex subunit E3; deltaD; deltaD transmembrane protein; delta-like protein D; diaphorase; dihydrolipoamide dehydrogenase; dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex); dihydrolipoamide: NAD+ oxidoreductase; dihydrolipoyl dehydrogenase mitochondrial-like protein; dihydrolipoyl dehydrogenase, mitochondrial; DLD; DLDD; DLDH; E3; E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex; GCSL; Glycine cleavage system L protein; glycine cleavage system protein L; LAD; lipoamide dehydrogenase; lipoamide dehydrogenase precursor; lipoamide reductase; lipoyl dehydrogenase; LOW QUALITY PROTEIN: dihydrolipoyl dehydrogenase, mitochondrial; mitochondrial Dihydrolipoyl dehydrogenase-like protein; OTTHUMP00000206744; OTTHUMP00000206746; OTTHUMP00000206748; OTTHUMP00000206749; PH; PHE 3; PHE3; QflA-16416; QtsA-11266; sc:d0402; unnamed protein product
Gene Symbols Dld
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13382, 1738, 298942
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.