Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLD Monoclonal Antibody (OTI6G6), TrueMAB™, OriGene
SDP

Catalog No. 502540920
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0920 30 μL
50-167-4323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0920 Supplier OriGene Supplier No. TA503421S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DLD Monoclonal antibody specifically detects DLD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen DLD
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6G6
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CCM2
Gene Accession No. Q9BSQ5
Gene Alias aei; after eight protein; branched chain alpha-keto acid dehydrogenase complex subunit E3; deltaD; deltaD transmembrane protein; delta-like protein D; diaphorase; dihydrolipoamide dehydrogenase; dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex); dihydrolipoamide: NAD+ oxidoreductase; dihydrolipoyl dehydrogenase mitochondrial-like protein; dihydrolipoyl dehydrogenase, mitochondrial; DLD; DLDD; DLDH; E3; E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex; GCSL; Glycine cleavage system L protein; glycine cleavage system protein L; LAD; lipoamide dehydrogenase; lipoamide dehydrogenase precursor; lipoamide reductase; lipoyl dehydrogenase; LOW QUALITY PROTEIN: dihydrolipoyl dehydrogenase, mitochondrial; mitochondrial Dihydrolipoyl dehydrogenase-like protein; OTTHUMP00000206744; OTTHUMP00000206746; OTTHUMP00000206748; OTTHUMP00000206749; PH; PHE 3; PHE3; QflA-16416; QtsA-11266; sc:d0402; unnamed protein product
Host Species Mouse
Immunogen Full length human recombinant protein of human CCM2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83605
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.