Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLEU7 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549518
Description

Specifications
DLEU7 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6UYE1 | |
DLEU7 | |
KLH conjugated synthetic peptide between 58-89 amino acids from human DLEU7. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DLEU7 | |
Leukemia-associated protein 7, Deleted in lymphocytic leukemia 7, DLEU7, LEU7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
220107 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction