Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP126611

Catalog No. NBP126611


Only null left
Add to Cart

Description

Description

DLG1 Polyclonal specifically detects DLG1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

DLG1
Polyclonal
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
discs, large homolog 1 (Drosophila), disks large homolog 1, DKFZp761P0818, DKFZp781B0426, hDlg, large (Drosophila) homolog 1, presynaptic protein SAP97
Rabbit
Affinity Purified
RUO
1739
Store at 4C. Do not freeze.
Western Blot, Immunoprecipitation
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 5-10 ug/mg lysate
Q12959
DLG1
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human synapse-associated protein 97 (discs, large homolog 1) using the numbering given in entry NP_004078.1 (GeneID 1739 ).
0.1 mL
Primary
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only