Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dlg1 Mouse anti-Rat, Alkaline Phosphatase, Clone: G28, Abnova™

Catalog No. 89338993 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-993 Supplier Abnova Corporation Supplier No. MAB19708

Mouse Monoclonal Antibody.

Dlg1 Monoclonal antibody specifically detects Dlg1 in Rat samples. It is validated for WB-Ti, IHC-P.

Specifications

Antigen Dlg1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone G28
Conjugate Alkaline Phosphatase (AP)
Description discs, large homolog 1 (Drosophila)
Gene Dlg1
Gene Alias Dlgh1, SAP97
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-104 of rat Dlg1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 25252
Target Species Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.