Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLG5 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128739V
Description
DLG5 Polyclonal antibody specifically detects DLG5 in Human samples. It is validated for Western BlotSpecifications
DLG5 | |
Polyclonal | |
Western Blot | |
discs, large homolog 5 (Drosophila), KIAA0583large (Drosophila) homolog 5, LP-DLG | |
The immunogen recognized by this antibody maps to a region between residue 1325 and 1375 of human discs, large homolog 5 using the numbering given in entry NP_004738.3 (GeneID 9231). | |
0.1 mL | |
Cell Cycle and Replication | |
9231 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction