Learn More
DLG5 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302302A

Description
The recommended shelf life for this product is 1 year from date of receipt.
May play a role at the plasma membrane in the maintece of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.Specifications
DLG5 | |
Polyclonal | |
Unconjugated | |
DLG5 | |
KIAA0583; PDLG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9231 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8TDM6 | |
DLG5 | |
Between 1325 and 1375 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.