Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLG5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | DLG5 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0405
![]() ![]() |
Proteintech Group Inc
156871AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
May play a role at the plasma membrane in the maintece of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.Specifications
DLG5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
E9Q9R9, Q8TDM6 | |
71228, 9231 | |
DLG5 Fusion Protein Ag8194 | |
Primary | |
-20°C | |
DLG5 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Discs large protein P dlg, Disks large homolog 5, DLG5, KIAA0583, LP DLG, P DLG5, PDLG, Placenta and prostate DLG | |
Dlg5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title