Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983038
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3038 Supplier Bioss Supplier No. BS13632R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role at the plasma membrane in the maintenance of the structure of epithelial cells and in the transmission of extracellular signals to the membrane and cytoskeleton.

Specifications

Antigen DLG5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Dlg5
Gene Accession No. Q8TDM6
Gene Alias 4933429D20Rik; discs large homolog 5; discs large MAGUK scaffold protein 5; discs large protein LP-DLG; Discs large protein P-dlg; discs, large homolog 5; discs, large homolog 5 (Drosophila); Disks large homolog 5; DLG5; KIAA0583; large type of P-DLG; LP-DLG; mKIAA0583; PDLG; P-DLG5; Placenta and prostate DLG; T25557
Gene Symbols Dlg5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DLG5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9231
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.