Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLG7/HURP Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP337984AF647
Description
DLG7/HURP Polyclonal antibody specifically detects DLG7/HURP in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
DLG7/HURP | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
Alexa Fluor 647 | |
DAP-5, Discs large homolog 7, discs, large (Drosophila) homolog-associated protein 5, disks large-associated protein 5, DLG7, Hepatoma up-regulated protein, HURPDLG1, KIAA0008discs, large homolog 7 (Drosophila), large homolog 7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 547-846 of human DLG7/HURP (NP_055565.3).,, Sequence:, KNVFRKKVVSGIASKPKQDDAGRIAARNRLAAIKNAMRERIRQEECAETAVSVIPKEVDKIVFDAGFFRVESPVKLFSGLSVSSEGPSQRLGTPKSVNKAVSQSRNEMGIPQQTTSPENAGPQNTKSEHVKKTLFLSIPESRSSIEDAQCPGLPDLIEENHVVNKTDLKVDCLSSERMSLPLLAGGVADDINTNKKEGISDVVEGMELNSSITSQDVLMSSPEKNTASQNSILEEGETKISQSELFDNKSLTTECHLLDSPGLNCSNPFTQLERRHQEHARHISFGGNLITFSPLQPGEF | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
9787 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction