Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLG7/HURP Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040572AF750
Description
DLG7/HURP Polyclonal antibody specifically detects DLG7/HURP in Human samples. It is validated for Western BlotSpecifications
DLG7/HURP | |
Polyclonal | |
Western Blot | |
DAP-5, Discs large homolog 7, discs, large (Drosophila) homolog-associated protein 5, disks large-associated protein 5, DLG7, Hepatoma up-regulated protein, HURPDLG1, KIAA0008discs, large homolog 7 (Drosophila), large homolog 7 | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of Human Hepatoma Upregulated Protein/Discs, Large 7 using the numbering given in entry NP_055565.2 (GeneID 9787). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
9787 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction