Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLG7/HURP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040572
Description
DLG7/HURP Polyclonal specifically detects DLG7/HURP in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
DLG7/HURP | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DAP-5, Discs large homolog 7, discs, large (Drosophila) homolog-associated protein 5, disks large-associated protein 5, DLG7, Hepatoma up-regulated protein, HURPDLG1, KIAA0008discs, large homolog 7 (Drosophila), large homolog 7 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, and Chimpanzee. | |
Store at 4C. Do not freeze. |
Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
NP_055565.2 | |
DLGAP5 | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of Human Hepatoma Upregulated Protein/Discs, Large 7 using the numbering given in entry NP_055565.2 (GeneID 9787). | |
100 μL | |
Core ESC Like Genes, Stem Cell Markers | |
9787 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction