Learn More
Invitrogen™ DLG7 Polyclonal Antibody

Description
Immunogen sequence: EYERNRHFGL KDVNIPTLEG RILVELDETS QELVPEKTNV KPRAMKTILG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | DLG7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | DLGAP5 |
| Gene Accession No. | Q15398 |
| Gene Alias | C77459; C86398; DAP-5; discs large homolog 7; discs large homolog associated protein 5; discs, large (Drosophila) homolog-associated protein 5; discs, large homolog 7; discs, large homolog-associated protein 5; disks large-associated protein 5; Disks large-associated protein DLG7; DLG associated protein 5; DLG7; DLGAP5; Hepatoma up-regulated protein; hepatoma up-regulated protein homolog; HURP; Kiaa0008; mKIAA0008 |
| Gene Symbols | DLGAP5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.