Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120226 Supplier Invitrogen™ Supplier No. PA5120226
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF-7, DU-145, K-562. The target is usually found in the following locations: Cytoplasm, Nucleus, cytoskeleton, spindle. Immunogen sequence: KNVFRKKVVS GIASKPKQDD AGRIAARNRL AAIKNAMRER IRQEECAETA VSVIPKEVDK IVFDAGFFRV ESPVKLFSGL SVSSEGPSQR LGTPKSVNKA VSQSRNEMGI PQQTTSPENA GPQNTKSEHV KKTLFLSIPE SRSSIEDAQC PGLPDLIEEN HVVNKTDLKV DCLSSERMSL PLLAGGVADD INTNKKEGIS DVVEGMELNS SITSQDVLMS SPEKNTASQN SILEEGETKI SQSELFDNKS LTTECHLLDS PGLNCSNPFT QLERRHQEHA RHISFGGNLI TFSPLQPGEF.

DLG7 is a potential cell cycle regulator that may play a role in carcinogenesis of cancer cells and mitotic phosphoprotein regulated by the ubiquitin-proteasome pathway. The protein is also a key regulator of adherens junction integrity and differentiation that may be involved in CDH1-mediated adhesion and signaling in epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DLG7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DLGAP5
Gene Accession No. Q15398, Q8K4R9
Gene Alias C77459; C86398; DAP-5; discs large homolog 7; discs large homolog associated protein 5; discs, large (Drosophila) homolog-associated protein 5; discs, large homolog 7; discs, large homolog-associated protein 5; disks large-associated protein 5; Disks large-associated protein DLG7; DLG associated protein 5; DLG7; DLGAP5; Hepatoma up-regulated protein; hepatoma up-regulated protein homolog; HURP; Kiaa0008; mKIAA0008
Gene Symbols DLGAP5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 547-846 of human DLGAP5 (NP_055565.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218977, 289997, 9787
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.