Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLGAP3 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305977FR
Description
DLGAP3 Polyclonal specifically detects DLGAP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DLGAP3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DAP-3, discs, large (Drosophila) homolog-associated protein 3, disks large-associated protein 3, SAP90/PSD 95 associated protein 3, SAP90/PSD-95-associated protein 3 | |
Produced in rabbits immunized with E. coli-derived Human DLGAP3 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
58512 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction