Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLGAP4 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176372PCP
Description
DLGAP4 Polyclonal antibody specifically detects DLGAP4 in Human samples. It is validated for Western Blot, ELISASpecifications
DLGAP4 | |
Polyclonal | |
Western Blot, ELISA | |
DAP4FLJ20588, discs, large (Drosophila) homolog-associated protein 4, disks large associated protein 4, disks large-associated protein 4, DLP4, KIAA0964MGC131862, PSD-95/SAP90-binding protein 4, RP5-977B1.6, SAP90/PSD-95-associated protein 4, SAPAP-4, SAPAP4DAP-4 | |
Antibody was raised against a 13 amino acid synthetic peptide from near the center of human SAPAP4. The immunogen is located within amino acids 610 - 660 of SAPAP4. Amino Acid Squence: APAPEAPEPPPKHA | |
0.1 mL | |
Signal Transduction | |
22839 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction