Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLGAP4 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298641PCP
Description
DLGAP4 Polyclonal antibody specifically detects DLGAP4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
DLGAP4 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
DAP4FLJ20588, discs, large (Drosophila) homolog-associated protein 4, disks large associated protein 4, disks large-associated protein 4, DLP4, KIAA0964MGC131862, PSD-95/SAP90-binding protein 4, RP5-977B1.6, SAP90/PSD-95-associated protein 4, SAPAP-4, SAPAP4DAP-4 | |
Produced in rabbits immunized with E. coli-derived Human DLGAP4 fragment. | |
0.1 mL | |
Signal Transduction | |
22839 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction