Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DLK1 Monoclonal Antibody (3A10)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA515915

Catalog No. PIMA515915


Only null left
Add to Cart

Description

Description

MA5-15915 targets DLK1 in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human samples. The MA5-15915 immunogen is purified recombinant fragment of human DLK1 expressed in E. Coli. . MA5-15915 detects DLK1 which has a predicted molecular weight of approximately 41kDa.

The Delta-like 1 homolog (DLK1) is a transmembrane protein containing six epidermal growth factor repeats. It is involved in the differentiation of several cell types, including adipocytes and is also thought to be a tumor suppressor. The DLK1 gene is one of several imprinted genes located in a region on chromosome 14q32; certain mutations in this imprinted region can cause phenotypes similar to maternal and paternal uniparental disomy of chromosome 14 (UPD14). DLK1 is expressed from the paternal allele; a polymorphism within this gene has been associated with child and adolescent obesity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DLK1
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
P80370, Q09163
DLK1
Purified recombinant fragment of human DLK1 expressed in E. Coli.
RUO
13386, 8788
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
3A10
Unconjugated
DLK1
Adipocyte differentiation inhibitor protein; AW742678; delta like non-canonical Notch ligand 1; Delta1; delta-like 1 homolog; delta-like 1 homolog (Drosophila); DLK; Dlk1; DLK-1; DlkI; FA1; Fetal antigen 1; Ly107; Peg9; pG2; Preadipocyte factor 1; preadipocyte factor-1; PREF; Pref1; pref-1; Protein delta homolog 1; SCP1; Scp-1; secredeltin; stromal cell derived protein 1; ZOG
Mouse
100 μL
Primary
Human, Mouse
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.