Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DLK1, Mouse anti-Human, Clone: PF13-3, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16055
Specifications
DLK1 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4. | |
DLK/FA1/PREF1/Pref-1/ZOG/pG2 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
PF13-3 | |
ELISA (1:2000-10000) Flow Cytometry (1:100-500) Immunohistochemistry (1:200-1000) Western Blot (1:2000-10000) The optimal working dilution should be determined by the end user. | |
delta-like 1 homolog (Drosophila) | |
DLK1 | |
Recombinant protein corresponding to extracellular domain of human DLK1. | |
RUO | |
8788 | |
For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction